Hub-Cap Adaptors, Restek
Fournisseur: Restek
Certificats
Certificats
A propos de cet article
Allow the use of the Mobi-Cap Top with 4-liter solvent bottles.
Customers Who Bought This Also Bought
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.9%, HiPerSolv CHROMANORM®, super gradient grade for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
$741.97 to $47,885.70
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
$57.66 to $576.72
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
$635.20 to $2,541.50