Pack in a Box Kit, Restek
# de catalogue 10853-808
Fournisseur: Restek
Spécifications
- Description:Pack in a Box Kit
- Cat. no.:10853-808
- Supplier No.:26408
Spécifications
A propos de cet article
Restek’s Pack in a Box kit is a complete column-packing system.
AC input requirement: 85 to 265 VAC. 10000 psi maximum pressure.
Delivery: Everything you need to pack your own analytical HPLC columns is included: pump, 20 ml reservoir, column hardware (150×4.6 mm, 316 stainless steel tubing with end-fittings, frits, nuts, and ferrules for two complete, empty columns), and system control software (computer not included). Detailed packing instructions and a DVD are also included. The packing pump is electric, so no pressurized air is required.
Customers Who Bought This Also Bought