Valves for HPLC, Restek
Fournisseur: Avantor
Certificats
A propos de cet article
Our valves have low internal volumes and are rated to 15,000 psi (103,421 kPa).
They feature a two-piece stem assembly in which the rotating upper shank is coupled through a ball joint to a static lower stem. Only finger-tight torque is required to make the seal.
Customers Who Bought This Also Bought