MiniPulse Pulse Dampeners, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Improve system baseline stability while increasing the total system volume by only 160 µl.
- Compact unit (2.5×1.5") can be placed almost anywhere
- Small, 160 µl dead volume at atmospheric pressure
- PEEK unit can withstand pressures to 5000 psi (34474 kPa)
- 316 stainless steel unit can withstand pressures to 6000 psi (41369 kPa)
The MiniPulse pulse dampeners are ideal for applications where minimizing the total system volume is critical. Stainless steel and PEEK options for a wide range of applications.
Customers Who Bought This Also Bought