PEEK Fitting Extractor, Restek
# de catalogue RK25325
Fournisseur: Restek
Spécifications
- Description:Tool, PEEK Fitting Extractor HPLC
- Cat. no.:RK25325
- Supplier No.:25325
Spécifications
A propos de cet article
Drill into the broken fitting, then screw the extractor into the fitting and remove it easily.
Eliminates the need for heat or other techniques that could damage your column.
Customers Who Bought This Also Bought