LO-Pulse Pulse Dampers, Restek
Fournisseur: Restek
Certificats
A propos de cet article
The LO-Pulse Pulse Dampers are patented, wide-dynamic-range 316 stainless steel device that smooths pulsations and maintains constant flow at system pressures from 500 to 6000 psi (3447 to 41369 kPa).
The flow path volume is only 0.9 ml and the path are efficiently swept, eliminating solvent memory effects when changing mobile phases.
Delivery: The LO-Pulse pulse damper kit version is a space-saving, economical kit that includes hardware for mounting the pulse damper on a bracket, or for installing feet on it for bench-top use.
Customers Who Bought This Also Bought