About this item
These syringes are used in chromatography and dissolution testing
- Safety stop to prevent accidental spills
- For situation needing an inert, non-reactive syringe
Designed with centered, Luer Lock tips, these syringes simply are the best for laboratory applications such as HPLC, Dissolution Testing, CE, LCMS.
They are latex free, contain no rubber, no silicone oil, styrene or DEHP.