VWR® Polystyrene Spot Plate
# de catalogue 470031-818
Fournisseur: CCMI, INC.
Spécifications
- Description:Polystyrene spot plate with 12 depressions
- Cat. no.:470031-818
- Supplier No.:2321028
Spécifications
A propos de cet article
Conduct your microchemistry experiments safely.
- Chemically inert
- Unbreakable
Not recommended for use with acetone, xylene, or other strong solvents.
Customers Who Bought This Also Bought