ContactUsLinkComponent
VWR® Polystyrene Spot Plate
VWR® Polystyrene Spot Plate
# de catalogue 470031-818
Fournisseur:  CCMI, INC.
SPOT PLATE POLYSTYRENE 12 WELLS
VWR® Polystyrene Spot Plate
# de catalogue 470031-818
Fournisseur:  CCMI, INC.
Spécifications
  • Description:
    Polystyrene spot plate with 12 depressions
  • Cat. no.:
    470031-818
  • Supplier No.:
    2321028
Spécifications
A propos de cet article
Conduct your microchemistry experiments safely.
  • Chemically inert
  • Unbreakable
Not recommended for use with acetone, xylene, or other strong solvents.
Customers Who Bought This Also Bought