Spec-Master® Series Stainless Steel Backsplash Work Table, 14-Gauge Type 300 Series, with Galvanized Steel Base and Undershelf, Eagle MHC™
# de catalogue 58202-434
Fournisseur: Eagle Group
Spécifications
- Description:E-BS Series Work Table
- Cat. no.:58202-434
- W×D:72×30"
- Supplier No.:T3072E-BS
Spécifications
A propos de cet article
14 Gauge, Type 300 series stainless steel top with 4½" backsplash at rear of table.
- Rolled edge front with ends turned under 90 degrees
- Adjustable working height (35⅛" to 36⅛")
1⅝" diameter legs and adjustable undershelf are heavy gauge galvanized steel.
Ordering information: Available accessories iinclude drawers and casters, sold separately.
Customers Who Bought This Also Bought