GravityFlo® Portable Eyewash, SE-4400, Speakman®
# de catalogue 10068-226
Fournisseur: Speakman
Spécifications
- Description:Gravityflo® portable eyewash
- Cat. no.:10068-226
- Supplier No.:SE-4400
Spécifications
A propos de cet article
The SE-4400 portable eyewash allows to locate an eyewash anywhere that it is needed, even where potable water is not available.
- Features a 34.06 L (9 gal.) tank, providing continuous flow
- Capability to be shelf, table, or wall mounted
- Pull down activation with hands free operation
- Plastic fill cap with integral tamper prevention and molded plastic sprays
Certifications: Meets ANSI/ISEA Z358.1.
Customers Who Bought This Also Bought