Monocot Stem Structure
# de catalogue 470220-636
Fournisseur: United Scientific Supplies
Spécifications
- Description:Monocot Stem Structure Model
- Height:17 cm
- Cat. no.:470220-636
- Length:45 cm
- Width:45 cm
- Supplier No.:MBMS01
- No. of Parts:1
Spécifications
A propos de cet article
Students can use this structure to examine a corn stem in cross and longitudinal sections.
- Model Displays Basic Structure of Monocot Stem
- Large Size for Easy Examination
- Detailed Key Included
- Durable Construction
- Injection-Molded Model
This model is patterned on a corn stem to show the general anatomy of a monocot in both longitudinal and cross sections. It reveals features such as the cuticle layer, epidermis, closed vascular bundles, and parenchyma cells.
This injection-molded model is more durable than the more common fiberglass construction.
Ordering information: This structure includes a detailed key.
Customers Who Bought This Also Bought