Rudiger® Physiological Spine
Physiological Spine
Catalog # 470221-152
Supplier: RUDIGER ANATOMIE GMBH
Fournisseur #: A207
Jump to:
Product Details & Documents
Lifelike accuracy cast from a natural spine.
- Fully articulated and flexible
- Soft discs compress like natural tissue
- 5 year manufacturers warranty
Our spine is fully articulated and flexible to show vertebral movements and alignments. Comes with arteries, spinal cord, nerve ends and flexible femur heads. The sacrum is sectioned to show course of spinal nerves. Contains a softer disc material, that compresses like natural tissue, bulging outward when under pressure. With stand.
Spécifications
- Désignation:Physiological Spine