Vileda® Professional Roll-O-Matic® Sponge Refills, Vileda Professional-FHP
Fournisseur: Vileda Professional - FHP
Certificats
A propos de cet article
The Vileda Professional Roll-O-Matic® sponge mop refills are manufactured in the USA
- Original refills have been the standard in the professional and controlled environment industry for well over 30 years
- Refills are designed for use in all Roll-O-Matic® applications
- Fully autoclavable
- Manufactured in the USA
All Original Refills are ideal for use in ISO 6, LAR, production, and laboratory environments.
All Vileda Professional brand refills are ideal for use with the "Pull and Lift" technique most commonly used in controlled environments. All 10" Roll-O-Matic® sponge refills fit every 10" Roll-O-Matic® handle; all 14" Roll-O-Matic® sponge refills fit every 14" Roll-O-Matic® handle. Certificate of conformance and technical data available upon request.
Customers Who Bought This Also Bought