7000 Series Reusable Half Mask Respirators, Pre-Assembled, Moldex®
Fournisseur: Moldex/Metric
Certificats
A propos de cet article
Oil-free or Oil/water based paint spray or pesticides pre-assembled respirators.
- 100% PVC-free
- NIOSH approved for organic vapors
- Oil-free or oil/water based
Areas of applications include degreasing, forging, foundries, heat treating operations, paper making, rendering, rubber manufacturing, manufacture and use of adhesives, dry cleaning, pesticide manufacture and application, treatment of timber and wood products, manufacture and use of inks, oil-based paint, printing, manufacture of cosmetic preparations, perfumes and pharmaceuticals, flour bleaching, agriculture, tobacco industry, mining, trucking, manufacture of roofing and asphalt products and petrochemical plants.
Spécifications
- Catalog No:
- 10065-334
- 10065-336
- 10065-332
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health