VWR® Double-Sided Tape
Supplier: VWR International
Certificates
About this item
Transparent double coated polyester film available in Hi and Lo Tack with liner.
- Double-coated
- Polyester fabrication
Hi Tack features a permanent solvent acrylic adhesive—easily bonds dissimilar surfaces.
Lo Tack features a permanent adhesive on one side and removable on the other—ideal for peel and re-seal applications.
Customers Who Bought This Also Bought