Magnesium Standards for ICP, Certipur®, MilliporeSigma
Fournisseur: MilliporeSigma
Certificats
A propos de cet article
Traceable to SRM from NIST.
Certifications: The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
Ordering information: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.