About this item
Extran® AP 18 is a universal cleaning agent for the gentle cleaning of alkali-sensitive items. Suitable for cleaning items which cannot be tainted with allergenic detergents, e.g. jewellery, glasses. Extran® AP 18 mild alkaline does not foam even during heavy agitation in a washing machine.
- Surfactant-free
- Mild alkaline liquid
- Chlorine-free
- Phosphate-free
- Free from odorants/dyestuffs
Extran® AP 18 mild alkaline is a liquid main cleaning agent with mild alkaline properties for automatic dosage.
Dosing
The ideal dosing depends on the hardness of the water and the level of contamination of the item to be cleaned.
Under normal conditions, the concentration for application is 0.3–0.5%, i.e. 30–50 mL of Extran® AP 18 are used for around 10 L of water
Sequestering agent content (complexometric) 33.0–36.0%
Free alkali (as ammonia) 3.0–4.0%
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- EM1.40118.9025
- EM1.40118.9010
- Formulation:
- Complexing agents, alkali salts
- Complexing agents, alkali salts