Eyesaver® Deck Plate Faucet and Eyewash Combination, Speakman®
Fournisseur: Speakman
Certificats
A propos de cet article
Eyesaver® faucet combines a sensor activated 8" deck plate gooseneck faucet with an independently operated pull-activated eyewash.
- Aerated yellow plastic spray outlets with flip-top dust caps
- Dedicated water channel for eyewash with pull handle activator
- ³/₈" compression connections with male threads for hot and cold inlets
Certifications: Meets ANSI/ISEA Z358.1, ASME A112.18.1/CSA B125.1.
Customers Who Bought This Also Bought