Panel-Mounted Valves, 45° Angle Face Fume Hood, with Four-Arm Handles, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
A propos de cet article
Panel mounted valve and outlet fitting for installation on angle face of hood.
- Concealed locking ring with threaded escutcheon and set screw
- Nylon four-arm handle with color-coded index disc
- 3/8" NPT female inlet and outlet
- Front load
Customers Who Bought This Also Bought