DIN 53217 Gravity Cup (Pyknometer), Industrial Physics
Fournisseur: GREAT LAKES GAGES, LLC
Certificats
A propos de cet article
The specific gravity cups are used for determining the specific gravity or density of coatings, pastes or similar liquids
Density is defined as weight per unit volume at specific temperature. These cups are avalable in anodised aluminium and stainless steel.
Certifications: Meets DIN 53217, ISO 2811, ASTM D 1475 standards.
Delivery: Inculdes Pyknometer and a calibration certificate.
Customers Who Bought This Also Bought