Order Entry
ContactUsLinkComponent
Shigella Broth Base, HiMedia
Shigella Broth Base, HiMedia
# de catalogue CA95020-940
Fournisseur:  HiMedia
Numéro CAS:  
Shigella Broth Base, HiMedia
# de catalogue CA95020-940
Fournisseur:  HiMedia
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Description:
    Shigella Broth
  • Size:
    500 g
  • Cat. no.:
    CA95020-940
  • Supplier No.:
    M1326-500G

Spécifications

A propos de cet article

Shigella Broth Base is used for the isolation and cultivation of Shigella species from food.

  • Contains casein enzymic hydrolysate as a source of carbon, nitrogen, vitamins and minerals
  • Buffering provided by dipotassium hydrogen phosphate and potassium dihydrogen phosphate
  • Sodium chloride maintains the osmotic balance of the medium

Shigella are gram-negative, non-motile, non-spore forming rod-shaped bacteria closely related to Escherichia coli and Salmonella. Shigella infection is typically via ingestion (faecal-oral contamination), depending on age and condition of the host, as few as ten bacterial cells can be enough to cause an infection. Shigella causes dysentery that results in the destruction of the epithelial cells of the intestinal mucosa in the cecum and rectum. Some strains produce enterotoxin and Shiga toxin, similar to the verotoxin of E. coli O157:H7. Shigella Broth Base is used for the isolation and cultivation of Shigella species, as recommended by APHA. Shigella Broth Base contains casein enzymic hydrolysate as a source of carbon, nitrogen, vitamins and minerals. Dextrose provides the necessary carbohydrates. Buffering action in the medium is provided by dipotassium hydrogen phosphate and potassium dihydrogen phosphate. Sodium chloride maintains the osmotic balance of the medium. Polysorbate 80 is inhibitory for growth of accompanying microflora besides providing growth factors. Novobiocin is inhibitory for gram-positive bacteria such as S. aureus and certain gram-negative organisms such as H. influenzae and some species of Proteus.

Customers Who Bought This Also Bought