Vileda® Professional Compact Bucket System
Fournisseur: Vileda Professional - FHP
Certificats
A propos de cet article
High performance in a compact design
- Perfect for use with all Roll-O-Matic® mops (without sieve)
- Solid and stable polypropylene plastic buckets and bails are 100% autoclavable
- Unique electro-polished sieve (sold separately) with hole pattern, integrated bucket splash guard, and handle support
The single bucket system includes one six gallon bucket and swivel plastic casters. Fully autoclavable, the casters are fixed to the bucket with a stainless steel pin connection. The double bucket system includes two six gallon buckets and an electro-polished stainless steel chasis that is designed to keep the buckets upright. Stainless steel autoclavable casters are fixed to the chasis with a stainless steel screw/nut connection. The electro-polished sieve is sold separately.
Cleanrooms are decreasing in size, affecting the areas to be cleaned as well as the storage rooms for cleaning equipment. The solution is the new Cleanroom Compact Bucket System concept.
Customers Who Bought This Also Bought