Dome Brush, Aluminum Ferrule, Justman Brush
# de catalogue 17145-027
Fournisseur: Justman Brush
Spécifications
- Bristle Color:Black
- Bristle Material:Goat hair
- Brush Head Diameter:19 mm (3/4")
- Total length:20.3 cm
- Cat. no.:17145-027
- Supplier No.:1924
- Brush Head Length:2.54 cm (1")
Spécifications
A propos de cet article
Dome brush is soft and static-resistant for delicate jobs
Useful for dusting balances.
Customers Who Bought This Also Bought