Oil Return Kit, Labconco
# de catalogue 75796-646
Fournisseur: Labconco
Spécifications
- Description:Oil Return Kit
- Cat. no.:75796-646
- Supplier No.:1473500
Spécifications
A propos de cet article
Adjustable Gas Ballast Oil Return Kit for field installation on Labconco Rotary Vane Vacuum Pumps (sold separately).
It returns oil from exhaust filter housing to pump. This is needed when pump is run with the gas ballast open for extended periods of time.
Ordering information: Kit includes one oil return assembly, one drain adaptor, one restrictor, one flexible oil-return tube, two hose clips, one ‘O’ ring: Ø 14.6×2.4 mm, one ‘O’ ring: Ø 21.5×3.0 mm, and installation instructions.
Customers Who Bought This Also Bought