Upchurch Scientific® Barbed Adapters, IDEX Health & Science
Fournisseur: Upchurch Scientific
Certificats
A propos de cet article
Barbed adapters are used to connect peristaltic-type flexible tubing for general, low pressure applications, such as plumbing peristaltic pumps.
- Made of polypropylene, class VI material
- Ideal for use with most aqueous solutions
- Can be sterilized via gamma radiation
Adapters feature an integrated locking ring surrounding the male luer nose for more secure connections. They are packaged individually and can be used with a variety of tubing inner diameters
Customers Who Bought This Also Bought