Spécifications
- Description:Loading Dye Base 50X
- Size:25 mL
- Supplier No.:1B1812-25ML
Spécifications
A propos de cet article
Loading Dye Base, 50X is used for custom loading buffer preparation.
- Custom loading buffer preparation
- Concentrated solution containing patent blue
- Eliminates the need to weigh messy dye powders
Loading Dye Base, 50X is used for custom loading buffer preparation. The Loading Dye Base is a concentrated solution containing patent blue, bromophenol blue and amaranth tracking dyes, which migrate at approximately 4,000 bp, 400 bp and 10 bp, on a 1% agarose gel, respectively. Using Loading Dye Base eliminates the need to weigh messy dye powders.