Wesco® Platform Trucks with Folding Handle
# de catalogue 21002-050
Fournisseur: Wesco
Spécifications
- Length:29"
- Width:19"
- Description:Platform Trucks with Steel Folding Handle
- Cat. no.:21002-050
- Shipping Weight:25 lbs.
- Load Capacity:440 lbs
- Finish:Enameled
- Supplier No.:272019
Spécifications
A propos de cet article
Steel Folding Handle Trucks feature fold-down handles for easy storage
- Protective vinyl perimeter bumpers
- 4" caster wheels made of TPR non-marking rubber
- UPS shippable
They have an enameled finish and a non-skid, heavy vinyl deck surface.
Customers Who Bought This Also Bought