Platinum 95%/Gold 5% Dishes, Heraeus
Supplier: HERAEUS MATERIALS TECHNOLOGY NA
Certificates
About this item
These dishes are made with 95% Platinum and 5% Gold.
Due to their resistance to oxidation and corrosion, as well as mechanical stability at high temperatures, Platinum Dishes have become essential for the preparation of samples in analytical laboratories.
The mechanical stability of Platinum is further improved with the addition of 5% Gold. This alloy offers higher strength and lower wettability, so that the sample can be easily removed from the dish.
Ordering information: Pourling Lip available upon request. Lids sold separately. For additional lid details, see Lids for Platinum and Platinum 95%/Gold 5% Dishes, Heraeus.
Customers Who Bought This Also Bought