PBT Screw Caps for DURAN® Laboratory Glass Bottles, Wheaton, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
The cap is available with a PTFE coated silicone seal or as a screw connection cap with aperture.
- Autoclavable
- Chemically resistant
- Temperature resistant to 180°C
Customers Who Bought This Also Bought