Respirator Storage Bags, Allegro
# de catalogue 47747-252
Fournisseur: Allegro
Small Business Enterprise
Spécifications
- Size:Large - 4 to 16"
- Cat. no.:47747-252
- Supplier No.:2000
Spécifications
A propos de cet article
Reusable storage bags keep equipment contaminant free.
- Ten Milliliter poly
- Transparent
- Reusable
- For full or half-mask sizes
These large storage bags are transparent and built with a zipper.
They're made of tough, 10 ml poly to last longer and hold half or full-mask sizes. Cleaning instructions are printed on the bags.
Customers Who Bought This Also Bought