Hand Crimper, PerkinElmer
# de catalogue CA97013-494
Fournisseur: PERKINELMER SCIENTIFIC CANADA ULC
Spécifications
- Description:Hand Crimper
- Cat. no.:CA97013-494
- Seal Size:20 mm
- Supplier No.:N9302785
Spécifications
A propos de cet article
Hand crimper for GC Headspace vials or LC large volume vials.
- Suited for large-volume vials
- Guarantees sample integrity
- Useful when handling volatile compounds
It ensures sample integrity, particularly those containing volatile compounds.
Customers Who Bought This Also Bought