Wind-Up Tape Measure
# de catalogue 470002-852
Fournisseur: Learning Resources
Spécifications
- Description:Metric/imperial ruler
- Material:Plastic
- Cat. no.:470002-852
- Length:10 m (33 ft.)
- Supplier No.:LER0365
Spécifications
A propos de cet article
Flexible Plastic Tape measures in metric and imperial units.
- Removable handle for easy control
- Durable hard plastic casing for years of use
Easy wind-up tapes are marked in inches and feet on one side and mm and cm on the other.
Customers Who Bought This Also Bought