About this item
The VWR® Wall Shelving System is perfect for storing items in spaces where order is paramount.
- Can be mounted directly or on a horizontal rail
- Powder-coated steel finish
- Reusable
- Can be dismantled and reinstalled with minimal effort
The shelving system can easily be modified according to need and is reusable, as it can be dismantled and reinstalled in a new location with minimal effort
The system allows quick and easy installation. Simply hang the vertical upright on the oval horizontal rail and slide to the desired width. Two horizontal rail widths are available, and two vertical upright lengths are available. Light neutral or snow white, powder-coated steel finish.
VWR® Wall Shelving can be mounted on the wall either directly or using a horizontal rail. The system consists of horizontal rails and upright profiles, with a wide range of steel shelves, as well as accessories (bin rails, cabinets, etc). Shelves are available in various standard widths and depths. All parts of the system are compatible.