ContactUsLinkComponent
TrueBlack Lipofuscin Autofluorescence Quencher, Biotium
TrueBlack Lipofuscin Autofluorescence Quencher, Biotium
# de catalogue 10119-144
Fournisseur:  Biotium
Numéro CAS:  
TrueBlack Lipofuscin Autofluorescence Quencher, Biotium
# de catalogue 10119-144
Fournisseur:  Biotium
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    TrueBlack Lipofuscin Autofluorescence Quencher, 20X in DMF
  • Size:
    1 mL
  • Cat. no.:
    10119-144
  • Supplier No.:
    23007
Spécifications
A propos de cet article
TrueBlack quenches autofluorescence from lipofuscin, allowing specific immunofluorescence staining.
  • Eliminates lipofuscin autofluorescence
  • Can reduce autofluorescence from non-lipofuscin sources
  • Has low red/far-red background compared to Sudan Black B
  • Can be used before or after immunostaining
  • Clears the way for fluorescence imaging of human and aged animal tissues
TrueBlack™ is a new reagent for quenching lipofuscin autofluorescence in tissue sections for immunofluorescence staining
Traditionally, Sudan Black B has been used to quench lipofuscin autofluorescence by incubating tissue sections with the dye after immunofluorescence staining. However, while it masks the autofluorescence from lipofuscin, Sudan Black B also introduces uniform non-specific background fluorescence in the red and far-red channels, limiting the use of fluorescent dyes in those wavelengths. Now Biotium has developed TrueBlack™ as a superior alternative to Sudan Black B. TrueBlack quenches lipofuscin fluorescence with far less increase in red/far-red background fluorescence. TrueBlack™ treatment can be performed before or after immunostaining. It is rapid, simple, and has minimal effect on signal from fluorescent antibodies or nuclear counterstains, thus preserving the signal-to-noise ratio of the immunostaining .
TrueBlack effectively eliminates lipofuscin autofluorescence in tissues like human brain and human retina. TrueBlack also can reduce autofluorescence from other sources, such as collagen, elastin, red blood cells, and general background fluorescence. It does not quench these sources of autofluorescence as completely as it does for lipofuscin, but it can substantially improve background in human and non-human tissue types.
Lipofuscin consists of highly autofluorescent granules of oxidized proteins and lipids that build up in the lysosomes of aging cells. Lipofuscin granules fluoresce brightly in all channels used for fluorescence microscopy, and accumulate in a variety of cell and tissue types with age. Consequently, imaging of specific immunofluorescence signal in certain adult human tissues or aged animal tissues can be virtually impossible unless methods are employed to quench or mask lipofuscin fluorescence.
Ordering information: TrueBlack™ is supplied as a 20X stock solution in DMF.
Customers Who Bought This Also Bought