Click-On In-Line Super Clean® Big Trap Filter Kits, Restek
Fournisseur: Restek
Certificats
A propos de cet article
'Quick connect' fittings with spring-loaded check valves allow filter replacement in seconds without tools or contamination.
- Largest-capacity traps available—run longer without replacing
- Stainless steel construction withstands pressures up to 1000 psig
- High-purity output ensures 99.9999% pure gas (maximum flow of 8 L/min)
Triple gas trap is ideal for purifying carrier gas.
Hydrocarbon trap removes hydrocarbon contamination from carrier gas to help eliminate ghost peaks.
Customers Who Bought This Also Bought