Helium-Specific Replacement Triple Trap, Restek
# de catalogue 10854-428
Fournisseur: Restek
Spécifications
- Description:Helium-Specific Replacement Triple Trap (removes oxygen, moisture and hydrocarbons)
- Cat. no.:10854-428
- Supplier No.:22473
Spécifications
A propos de cet article
This helium-specific triple trap effectively removes oxygen, moisture, and hydrocarbons from GCs.
Customers Who Bought This Also Bought