Carbon Refill for Refillable Hydrocarbon Trap, Restek
# de catalogue RK20626
Fournisseur: Restek
Spécifications
- Description:Carbon Refill (two recharges)
- Cat. no.:RK20626
- Supplier No.:20626
Spécifications
A propos de cet article
Ideal for use with purge-and-trap systems to stop carrier gas interferences.
Remove trace organic impurities from carrier gas to 0.1 ppm where input does not exceed 100 ppm.
Customers Who Bought This Also Bought