Replacement Parts and Accessories for Mininert® Precision Sampling Valves for Vials, Restek
Catalog # 10854-244
Supplier: Restek
Specifications
- Description:Septum Insertion Tool
- Cat. no.:10854-244
- Supplier No.:24907
Specifications
About this item
These replacement parts for Mininert® Precision sampling valves for vials are extremely functional and economical.
- Replacement septa are delivered in packs of 50
- Septum insertion tool is made of durable materials
For use with Mininert® Precision sampling valves for vials.
Customers Who Bought This Also Bought