Capillary Column Rinsing Reservoir, Reservoir only, Restek
Supplier: Restek
Certificates
About this item
Rinsing reservoirs can be filled with solvent rinses and are ideal for cleaning capillary columns.
Restore the performance of bonded-phase capillary columns by dissolving and removing soluble, nonvolatile residue using this reservoir kit.
The 50 mL rinsing reservoir is equipped with 1/4 " inlet and outlet connections and includes a built-in fritted disk to prevent particulate matter from contaminating the column. The kit includes reservoir, pressure regulator, fittings, ferrules, and tubing. Reservoir also available separately.
Delivery: Column not included.
Customers Who Bought This Also Bought