Standard Ferrules for PerkinElmer GCs, Fit ≤0.28 mm int.Ø Fused Silica Columns, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Standard ferrules fit fused silica columns with internal diameters ≤0.28 mm.
- VG2, 60% Vespel®/40% graphite blend offers the best combination of sealing and ease of workability
- VG1, 85% Vespel®/15% graphite blend offers equivalent composition to Agilent Vespel®/graphite ferrules
- Seal with minimal torque, reusable, and preferred for vacuum and high-pressure uses
- Stable to 400°C
- Recommended for mass spec transfer lines
Customers Who Bought This Also Bought