Replacement Inlet Seals for Thermo TRACE 1300/1310 GCs, Restek
Fournisseur: Restek
Certificats
A propos de cet article
The inlet seal at the base of the GC injection port contacts the sample and must be changed frequently to prevent adsorption of active compounds.
- All seals include washers
In addition, septum fragments and sample residue accumulate on the disk surface, requiring disk replacement.
BodyTextAccessories: Note: The 1.2 mm inlet seal is recommended when installing two columns using a two-hole Vespel®/graphite ferrule.
Customers Who Bought This Also Bought