Flip Seal® Dual Vespel® Ring Inlet Seals, Restek
Fournisseur: Restek
Certificats
A propos de cet article
The reversible, two-sided design of these ring inlet seals allow significantly more analyses than other seals.
- Vespel ring embedded in top and bottom surface eliminates need for a washer
- Very little torque required to make seal
Highly inert gold or Siltek®-deactivated seals reduce breakdown and adsorption of active compounds, maximizing component transfer to GC column.
Ordering information: Note: The Flip Seal® inlet seal requires a special reducing nut adaptor fitting, which is included in the kit.
The Flip Seal® Adaptor can be used with standard 1/16" ferrules.
The Flip Seal® Adaptor can be used with standard 1/16" ferrules.
Customers Who Bought This Also Bought