Ferrules for Micropacked Columns, Capillary—for ¹/₁₆" Compression-Type Fittings, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Ferrules are designed for use with micropacked columns, with a capillary design ideal for 1/16" compression-type fittings
- Graphite construction
- 1/16" fitting size
Fit columns with internal diameters of 0.75 mm.
Ordering information: For micropacked columns.
Customers Who Bought This Also Bought