Ferrules for Thermo Scientific GCs, Fit ≤0.28 mm int.Ø Fused Silica Columns, Restek
Fournisseur: Restek
Certificats
A propos de cet article
For use with Thermo Scientific GCs
- Preconditioned to eliminate outgassing
- High-purity, high-density graphite
- Stable to 450°C
These ferrules contain no binders that can off-gas or adsorb analytes. Suitable for columns with internal diameters of 0.18–0.28 mm.
Customers Who Bought This Also Bought