Compact Ferrules for Agilent GCs, Fit 0.53 mm int.Ø MXT® Columns, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Compact ferrules are ideal for use with Agilent GCs.
- Durable, leak-tight
- Preconditioned to eliminate out-gassing
- Seal with minimal torque, reusable, and preferred for vacuum and high-pressure uses
- Recommended for mass spec transfer lines
Ferrules feature internal diameters of 0.8 mm, compatible with columns 0.45 or 0.53 mm in diameter.
Customers Who Bought This Also Bought