Packed Column Inlet Adapter Kit for ¹/₄" Injection Ports, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Packed column inlet adapter kits center columns perfectly in injection ports to eliminate bent syringe needles.
- Slotted design prevents carrier gas occlusion
- Vespel/graphite-reducing ferrules make installation easy
- Use 1/8" OD columns in 1/4" on-column injection ports
Delivery: Adapter kits includes all nuts and ferrules used to attach tubing to the injector or detector.
Customers Who Bought This Also Bought