Click-On In-line Super Clean® Ultra-High Capacity Moisture Trap, Restek
# de catalogue 10855-368
Fournisseur: Restek
Spécifications
- Description:Ultra-High Capacity Moisture Trap
- Cat. no.:10855-368
- Supplier No.:22467
Spécifications
A propos de cet article
Click-on traps measure 21.6×3.2 cm (8.5×1.25").
- Effective trap for catching moisture
Ordering information: Replacement trap only - fittings sold separately.
Customers Who Bought This Also Bought