Click-On In-line Super Clean® Ultra-High Capacity Hydrocarbon Trap, Restek
Catalog # 10855-236
Supplier: Restek
Specifications
- Description:Ultra-High Capacity Hydrocarbon Trap
- Cat. no.:10855-236
- Supplier No.:22466
Specifications
About this item
Click-on traps measure 21.6×3.2 cm (8.5×1.25").
- Effective trap for catching hydrocarbons
Ordering information: Replacement trap only- fittings sold separately.
Customers Who Bought This Also Bought