Ball Valve, Swagelok® Shut-Off Gas Valve, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
- Fittings are available in brass or stainless steel
Ball valves provide 1/4 turn for on-off control of fluids utilized in process and instrumentation applications. Available with connection sizes of 1/8" or 1/4" in diameter.
Customers Who Bought This Also Bought