Nut and Ferrule Set, Swagelok®, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
- Fittings are available in brass or stainless steel
No coating required, but nuts and ferrules can be treated upon request (custom parts).
Ordering information: Note: Nuts and ferrules are not treated unless requested (custom parts). Nuts and ferrules normally are not in contact with sample pathway, and thus do not require coating.
Customers Who Bought This Also Bought